2-Aminobenzo[d]thiazole-7-carboxylic acid 97%

Lieferant: Apollo Scientific

OR480523-250MG
APOSOR480523-250MGEA 125 EUR
APOSOR480523-250MG
2-Aminobenzo[d]thiazole-7-carboxylic acid 97%
2-Aminobenzo[d]thiazole-7-carboxylic acid
Formel: C₈H₆N₂O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD13195418
CAS-Nummer: 71224-95-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR