Ethyl-2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate 97%

Lieferant: Apollo Scientific

Synonyme: 2-Amino-5,6-dihydro-4-[4H]-cyclopenta[B]thiophene-3-carboxylic acid ethyl ester, Ethyl 2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate

OR480524-25G
APOSOR480524-25GEA 114 EUR
APOSOR480524-25G
Ethyl-2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate 97%
Ethyl-2-amino-5,6-dihydro-4H-cyclopenta[b]thiophene-3-carboxylate
Formel: C₁₀H₁₃NO₂S
Molecular Weight: 211.28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00102063
CAS-Nummer: 4815-29-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR