5-Nitrobenzo[d]thiazol-2-amine 95+%

Lieferant: Apollo Scientific

OR480528-1G
APOSOR480528-1GEA 63.8 EUR
APOSOR480528-1G
5-Nitrobenzo[d]thiazol-2-amine 95+%
5-Nitrobenzo[d]thiazol-2-amine
Formel: C7H5N3O2S
Lagertemperatur: Raumtemperatur
MDL: MFCD00160073
CAS-Nummer: 73458-39-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR