4-Bromo-2-chlorobenzo[d]thiazole 97%

Lieferant: Apollo Scientific

OR480529-1G
APOSOR480529-1GEA 106 EUR
APOSOR480529-1G
4-Bromo-2-chlorobenzo[d]thiazole 97%
4-Bromo-2-chlorobenzo[d]thiazole
Formel: C₇H₃BrClNS
Lagertemperatur: Raumtemperatur
MDL: MFCD04971817
CAS-Nummer: 182344-57-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR