4-Amino-2-(1,3-benzothiazol-2-yl)phenol 97%

Lieferant: Apollo Scientific

Synonyme: 4-Amino-2-(benzo[d]thiazol-2-yl)phenol

OR480541-250MG
APOSOR480541-250MGEA 143 EUR
APOSOR480541-250MG
4-Amino-2-(1,3-benzothiazol-2-yl)phenol 97%
4-Amino-2-(1,3-benzothiazol-2-yl)phenol
Formel: C₁₃H₁₀N₂OS
Schmelzpunkt: 184…190 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00197455
CAS-Nummer: 30616-38-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR