2-Amino-3-bromo-6-chloropyrazine

Lieferant: Apollo Scientific

OR480567-5G
APOSOR480567-5GEA 78.3 EUR
APOSOR480567-5G
2-Amino-3-bromo-6-chloropyrazine
2-Amino-3-bromo-6-chloropyrazine
Formel: C₄H₃BrClN₃
Lagertemperatur: Raumtemperatur
MDL: MFCD11044158
CAS-Nummer: 212779-21-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR