Methyl-2-methylnicotinat 97%

Lieferant: Apollo Scientific

Synonyme: methyl 2-methylpyridine-3-carboxylate, Methyl 2-methylnicotinate

OR480568-25G
APOSOR480568-25GEA 78.3 EUR
APOSOR480568-25G
Methyl-2-methylnicotinat 97%
Methyl-2-methylnicotinat
Formel: C₈H₉NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00051961
CAS-Nummer: 65719-09-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR