2-Chlorethyltoluol-4-sulfonat 97%

Lieferant: Apollo Scientific

Synonyme: 2-Chloroethyl p-toluenesulphonate

OR480570-100G
APOSOR480570-100GEA 162 EUR
APOSOR480570-100G
2-Chlorethyltoluol-4-sulfonat 97%
2-Chlorethyltoluol-4-sulfonat
Formel: C₉H₁₁ClO₃S
Molecular Weight: 234.7 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00000970
CAS-Nummer: 80-41-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR