N-(tert-Butoxycarbonyl)-D-2-phenylglycine 98%

Lieferant: Apollo Scientific

Synonyme: N-Boc-D-2-phenylglycine, BOC-D-alpha-phenylglycine, Boc-D-Phg-OH

OR480572-100G
APOSOR480572-100GEA 75.4 EUR
APOSOR480572-100G
N-(tert-Butoxycarbonyl)-D-2-phenylglycine 98%
N-(tert-Butoxycarbonyl)-D-2-phenylglycine
Formel: C₁₃H₁₇NO₄
Molecular Weight: 251,28 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00062043
CAS-Nummer: 33125-05-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR