(R)-Methyl-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride 98%

Lieferant: Apollo Scientific

Synonyme: H-D-Trp-OMe·HCl, D-Tryptophan methyl ester hydrochloride, H-D-Trp-OMe.HCl

OR480575-500G
APOSOR480575-500GEA 221 EUR
APOSOR480575-500G
(R)-Methyl-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride 98%
(R)-Methyl-2-amino-3-(1H-indol-3-yl)propanoate hydrochloride
Formel: C₁₂H₁₄N₂O₂·HCl
Molecular Weight: 254,7 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00038992
CAS-Nummer: 14907-27-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR