5-Amino-2-mercaptobenzimidazol 96%

Lieferant: Apollo Scientific

Synonyme: 5-Amino-1H-benzoimidazole-2-thiol

OR480589-100G
APOSOR480589-100GEA 287 EUR
APOSOR480589-100G
5-Amino-2-mercaptobenzimidazol 96%
5-Amino-2-mercaptobenzimidazole
Formel: C₇H₇N₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD00022671
CAS-Nummer: 2818-66-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR