2,4-Dimethylbenzolsulfonylchlorid 97%

Lieferant: Apollo Scientific

OR480597-100G
APOSOR480597-100GEA 0 EUR
APOSOR480597-100G
2,4-Dimethylbenzolsulfonylchlorid 97%
2,4-Dimethylbenzolsulfonylchlorid
Formel: C₈H₉ClO₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD00156143
CAS-Nummer: 609-60-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR