(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride 97%

Lieferant: Apollo Scientific

OR480598-100G
APOSOR480598-100GEA 0 EUR
APOSOR480598-100G
(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride 97%
(R)-Methyl-2-amino-3-hydroxypropanoate hydrochloride
Formel: C₄H₉O₃N·HCl
Lagertemperatur: Raumtemperatur
MDL: MFCD00066121
CAS-Nummer: 5874-57-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR