Isochinolin-6-carbonitril
Lieferant: Apollo Scientific
|
Achtung
|
OR48060-250MG
APOSOR48060-250MGEA
43.7
EUR
APOSOR48060-250MG
Isochinolin-6-carbonitril
Isochinolin-6-carbonitril
Formel:
C₁₀H₆N₂ Molecular Weight: 154,1680 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD09991660 CAS-Nummer: 106778-42-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...