Phenyl N-methylcarbamate 97%

Lieferant: Apollo Scientific

Synonyme: Phenyl methylcarbamate

OR480603-100G
APOSOR480603-100GEA 2140 EUR
APOSOR480603-100G
Phenyl N-methylcarbamate 97%
Phenyl N-methylcarbamate
Formel: C₈H₉NO₂
Molecular Weight: 151,163 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD01738783
CAS-Nummer: 1943-79-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR