N-Cbz-4-hydroxycyclohexane 97%

Lieferant: Apollo Scientific

OR480604-25G
APOSOR480604-25GEA 613 EUR
APOSOR480604-25G
N-Cbz-4-hydroxycyclohexane 97%
N-Cbz-4-hydroxycyclohexane
Formel: C₁₄H₁₉NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD02728352
CAS-Nummer: 16801-62-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR