Diethylterephthalat
Lieferant: Apollo Scientific
Synonyme:
Diethyl benzene-1,4-dicarboxylate
OR480605-100G
APOSOR480605-100GEA
132
EUR
APOSOR480605-100G
Diethylterephthalat
Diethylterephthalat
Formel:
C₁₂H₁₄O₄ Molecular Weight: 222,24 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00039891 CAS-Nummer: 636-09-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...