2,4,5-Trichlorbenzolsulfonamid

Lieferant: Apollo Scientific

Synonyme: 2,4,5-Trichlorbenzolsulfonsäureamid

OR480612-25G
APOSOR480612-25GEA 249 EUR
APOSOR480612-25G
2,4,5-Trichlorbenzolsulfonamid
2,4,5-Trichlorbenzolsulfonamid
Formel: C₆H₄Cl₃NO₂S
Molecular Weight: 260,53 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00119361
CAS-Nummer: 29092-27-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR