6-Chlor-1,3-dimethyluracil 98%

Lieferant: Apollo Scientific

OR480621-25G
APOSOR480621-25GEA 77 EUR
APOSOR480621-25G
6-Chlor-1,3-dimethyluracil 98%
6-Chlor-1,3-dimethyluracil
Formel: C₆H₇ClN₂O₂
Schmelzpunkt: 109…113 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00038066
CAS-Nummer: 6972-27-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR