1-Ethoxycarbonylcyclopropanecarboxylic acid 97%

Lieferant: Apollo Scientific
Achtung

OR480622-5G
APOSOR480622-5GEA 55.1 EUR
APOSOR480622-5G
1-Ethoxycarbonylcyclopropanecarboxylic acid 97%
1-Ethoxycarbonylcyclopropanecarboxylic acid
Formel: C₇H₁₀O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD08690114
CAS-Nummer: 3697-66-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR