(4-Iodphenyl)essigsäure 95%
Lieferant: Apollo Scientific
|
Achtung
|
OR480627-100G
OR480627-25G
APOSOR480627-100GEA
246
EUR
APOSOR480627-100G
APOSOR480627-25G
(4-Iodphenyl)essigsäure 95%
(4-Iodphenyl)essigsäure
Formel:
(C₆H₄I)CH₂COOH Schmelzpunkt: 136…140 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00082985 CAS-Nummer: 1798-06-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...