L(-)-Kampfer 97%

Lieferant: Apollo Scientific
Achtung

Synonyme: (1S)-(-)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanon, (1S,4S)-(-)-1,7,7-Trimethylbicyclo[2.2.1]-2-heptanon

OR480628-100G
APOSOR480628-100GEA 621 EUR
APOSOR480628-100G
L(-)-Kampfer 97%
L(-)-Kampfer
Formel: C₁₀H₁₆O
Molecular Weight: 152,24 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00064148
CAS-Nummer: 464-48-2
UN: 2717
ADR: 4.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR