N-(1-Naphthyl)ethylenediamine Dihydrochlorid 92%

Lieferant: Apollo Scientific
Achtung

Synonyme: N-(1-Naphthyl)ethylendiamin Dihydrochlorid, N-(1-Naphthyl)ethylendiammoniumdichlorid, Marshall Reagenz, N-2-Aminoethyl-1-naphthylamin Dihydrochlorid, 2-((Aminoethyl)amino)naphthalin Dihydrochlorid

OR480629-100G OR480629-5G OR480629-25G
APOSOR480629-100GEA 0 EUR
APOSOR480629-100G APOSOR480629-5G APOSOR480629-25G
N-(1-Naphthyl)ethylenediamine Dihydrochlorid 92%
N-2-Aminoethyl-1-naphthylamin Dihydrochlorid
Formel: C₁₂H₁₆Cl₂N₂
Molecular Weight: 259.18 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00012556
CAS-Nummer: 1465-25-4
Merck Index: 12,06496

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR