Ethyl-N-benzoyl-L-tyrosinat 98%
Lieferant: Apollo Scientific
Synonyme:
BTEE, Bz-Tyr-OEt
OR480630-100G
APOSOR480630-100GEA
331
EUR
APOSOR480630-100G
Ethyl-N-benzoyl-L-tyrosinat 98%
Ethyl-N-benzoyl-L-tyrosinat
Formel:
C₁₈H₁₉NO₄ Molecular Weight: 313,35 g/mol Lagertemperatur: Kühlschrank |
MDL:
MFCD00002388 CAS-Nummer: 3483-82-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...