2-(Carbobenzoxyamino)-1-ethanol 98%

Lieferant: Apollo Scientific

OR480631-100G
APOSOR480631-100GEA 87 EUR
APOSOR480631-100G
2-(Carbobenzoxyamino)-1-ethanol 98%
2-(Carbobenzoxyamino)-1-ethanol
Formel: C₁₀H₁₃NO₃
Lagertemperatur: Raumtemperatur
MDL: MFCD00191060
CAS-Nummer: 77987-49-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR