N-Fmoc-L-norleucin 98%

Lieferant: Apollo Scientific

OR480632-25G
APOSOR480632-25GEA 0 EUR
APOSOR480632-25G
N-Fmoc-L-norleucin 98%
N-Fmoc-L-norleucin
Formel: C₂₁H₂₃NO₄
Lagertemperatur: Raumtemperatur
MDL: MFCD00037537
CAS-Nummer: 77284-32-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR