6-Methoxy-2-naphthylglyoxal hydrate 97%

Lieferant: Apollo Scientific

OR480635-5G
APOSOR480635-5GEA 0 EUR
APOSOR480635-5G
6-Methoxy-2-naphthylglyoxal hydrate 97%
6-Methoxy-2-naphthylglyoxal hydrate
Formel: C₁₃H₁₂O₄
Lagertemperatur: Raumtemperatur
MDL: MFCD05664091
CAS-Nummer: 745783-88-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR