1-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate 98%

Lieferant: Apollo Scientific

Synonyme: 1-Benzyl N-Boc-L-aspartate, N-(tert-Butoxycarbonyl)-L-aspartic Acid 1-Benzyl Ester, BOC-L-Aspartic acid 1-benzyl ester, N-Boc-L-aspartic Acid 1-Benzyl Ester, Boc-Asp-OBzl, Boc-Asp-Obzl

OR480637-25G OR480637-100G
APOSOR480637-25GEA 46.4 EUR
APOSOR480637-25G APOSOR480637-100G
1-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate 98%
1-Benzyl-N-(tert-butoxycarbonyl)-L-aspartate
Formel: C₁₆H₂₁NO₆
Schmelzpunkt: 96…100 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00065563
CAS-Nummer: 30925-18-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR