2-Iod-5-nitrotoluol 97%

Lieferant: Apollo Scientific

Synonyme: 1-Iodo-2-methyl-4-nitrobenzene

OR480645-100G
APOSOR480645-100GEA 142 EUR
APOSOR480645-100G
2-Iod-5-nitrotoluol 97%
2-Iod-5-nitrotoluol
Formel: C₇H₆INO₂
Molecular Weight: 263.03 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00051517
CAS-Nummer: 5326-38-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR