2,6-Dihydroxynaphthalin 97%

Lieferant: Apollo Scientific

Synonyme: 2,6-Naphthalindiol

OR480646-25G
APOSOR480646-25GEA 132 EUR
APOSOR480646-25G
2,6-Dihydroxynaphthalin 97%
2,6-Dihydroxynaphthalin
Formel: C₁₀H₈O₂
Molecular Weight: 160,17 g/mol
Schmelzpunkt: 220 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00004082
CAS-Nummer: 581-43-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR