Cyclopentyl thiocyanate 95%
Lieferant: Apollo Scientific
Synonyme:
Cyclopentanethiocyanate, Cyclopentylrhodanid
OR480649-250G
APOSOR480649-250GEA
139
EUR
APOSOR480649-250G
Cyclopentyl thiocyanate 95%
Cyclopentyl thiocyanate
Formel:
C₆H₉NS Molecular Weight: 127,21 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD11100558 CAS-Nummer: 5263-57-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...