1-(4-Methylphenyl)cyclopropancarbonsäure 95%

Lieferant: Apollo Scientific

Synonyme: 1-(p-Tolyl)cyclopropanecarboxylic acid

OR480650-5G
APOSOR480650-5GEA 236 EUR
APOSOR480650-5G
1-(4-Methylphenyl)cyclopropancarbonsäure 95%
1-(4-Methylphenyl)cyclopropanecarboxylic acid
Formel: C₁₁H₁₂O₂
Molecular Weight: 176.2100 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00066918
CAS-Nummer: 83846-66-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR