Benzyl-acrylat 97%

Lieferant: Apollo Scientific

OR480651-100G
APOSOR480651-100GEA 0 EUR
APOSOR480651-100G
Benzyl-acrylat 97%
Benzyl acrylate
Formel: C₁₀H₁₀O₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00048147
CAS-Nummer: 2495-35-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR