(±)-1,2,3,4-Tetrahydro-1-naphthylamin 95%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
Tetralin-1-amine, 1,2,3,4-Tetrahydronaphthalen-1-amine
OR480652-25G
APOSOR480652-25GEA
89.9
EUR
APOSOR480652-25G
(±)-1,2,3,4-Tetrahydro-1-naphthylamin 95%
(±)-1,2,3,4-Tetrahydro-1-naphthylamin
Formel:
C₁₀H₁₃N Molecular Weight: 147,22 g/mol Siedepunkt: 246…247 °C/714mmHg Dichte: 1,026 g/cm³ (25 °C) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001740 CAS-Nummer: 2217-40-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...