(1S,2R)-cis-2-Aminocyclohexanol Hydrochlorid 98%

Lieferant: Apollo Scientific

OR480654-1G OR480654-5G
APOSOR480654-1GEA 0 EUR
APOSOR480654-1G APOSOR480654-5G
(1S,2R)-cis-2-Aminocyclohexanol Hydrochlorid 98%
(1S,2R)-cis-2-Aminocyclohexanol Hydrochlorid
Formel: C₆H₁₄ClNO
Lagertemperatur: Raumtemperatur
MDL: MFCD11618003
CAS-Nummer: 200352-28-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR