Ethyl (E)-3-(1-pyrrolidinyl)crotonate 97%

Lieferant: Apollo Scientific

Synonyme: (E)-Ethyl 3-(pyrrolidin-1-yl)but-2-enoate

OR480655-25G
APOSOR480655-25GEA 82.8 EUR
APOSOR480655-25G
Ethyl (E)-3-(1-pyrrolidinyl)crotonate 97%
Ethyl (E)-3-(1-pyrrolidinyl)crotonate
Formel: C₁₀H₁₇NO₂
Lagertemperatur: Raumtemperatur
MDL: MFCD00014097
CAS-Nummer: 54716-02-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR