Tetrachlorcyclopropen 98%

Lieferant: Apollo Scientific

OR480662-25G OR480662-5G
APOSOR480662-25GEA 446 EUR
APOSOR480662-25G APOSOR480662-5G
Tetrachlorcyclopropen 98%
Tetrachlorocyclopropene
Formel: C₃Cl₄
Lagertemperatur: Raumtemperatur
CAS-Nummer: 6262-42-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR