(2S)-2-Bromo-3-phenylpropanoic acid 96%

Lieferant: Apollo Scientific
Achtung

Synonyme: (S)-2-Bromo-3-phenylpropionic acid

OR480663-1G
APOSOR480663-1GEA 127 EUR
APOSOR480663-1G
(2S)-2-Bromo-3-phenylpropanoic acid 96%
(2S)-2-Bromo-3-phenylpropanoic acid
Formel: C₉H₉BrO₂
Molecular Weight: 229.071 g/mol
Lagertemperatur: Kühlschrank
MDL: MFCD00210121
CAS-Nummer: 35016-63-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR