gamma-Phenyl-gamma-Butyrolacton 99%
Lieferant: Apollo Scientific
|
Achtung
|
Synonyme:
5-Phenyltetrahydrofuran-2-one
OR480667-25G
APOSOR480667-25GEA
203
EUR
APOSOR480667-25G
gamma-Phenyl-gamma-Butyrolacton 99%
gamma-Phenyl-gamma-butyrolactone
Formel:
C₁₀H₁₀O₂ Molecular Weight: 162,19 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00005398 CAS-Nummer: 1008-76-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...