BOC-L-phenylalanine-N-hydroxysuccinimide ester 98%

Lieferant: Apollo Scientific

Synonyme: Boc-Phe-OSu

OR480671-25G
APOSOR480671-25GEA 85.7 EUR
APOSOR480671-25G
BOC-L-phenylalanine-N-hydroxysuccinimide ester 98%
BOC-L-phenylalanine-N-hydroxysuccinimide ester
Formel: C₁₈H₂₂N₂O₆
Molecular Weight: 362,382 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00037911
CAS-Nummer: 3674-06-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR