2-Nitrobenzoylchlorid 95%

Lieferant: Apollo Scientific

OR480675-100G
APOSOR480675-100GEA 1010 EUR
APOSOR480675-100G
2-Nitrobenzoylchlorid 95%
2-Nitrobenzoylchlorid
Formel: C₇H₄ClNO₃
Molecular Weight: 185,57 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD00007131
CAS-Nummer: 610-14-0
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR