2-Isocyanato-1-methoxy-4-methylbenzene 98%

Lieferant: Apollo Scientific

Synonyme: 2-Methoxy-5-methylphenyl isocyanate, 2-Methoxy-5-methylphenylisocyanate

OR480677-25G
APOSOR480677-25GEA 256 EUR
APOSOR480677-25G
2-Isocyanato-1-methoxy-4-methylbenzene 98%
2-Isocyanato-1-methoxy-4-methylbenzene

When melted, may contain small amount of sediment.

Formel: C₉H₉NO₂
Molecular Weight: 163,18 g/mol
Siedepunkt: 240 °C/760mmHg
Schmelzpunkt: 25…26 °C
Dichte: 1,112 g/cm³ (25 °C)
Lagertemperatur: Gefrierschrank
MDL: MFCD00013869
CAS-Nummer: 59741-04-7
UN: 2206
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR