6-Brom-2-hydroxypyridin 97%
Lieferant: Apollo Scientific
Synonyme:
2-Brom-6-pyridinol, 6-Brom-2-pyridinol, 2-Brom-6-hydroxypyridin
OR480679-25G
APOSOR480679-25GEA
59.6
EUR
APOSOR480679-25G
6-Brom-2-hydroxypyridin 97%
6-Brom-2-hydroxypyridin
Formel:
C₅H₄BrNO Molecular Weight: 174 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00234043 CAS-Nummer: 27992-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...