4-Phenoxybenzenesulphonamide 95%

Lieferant: Apollo Scientific

OR480681-25G OR480681-5G
APOSOR480681-25GEA 1010 EUR
APOSOR480681-25G APOSOR480681-5G
4-Phenoxybenzenesulphonamide 95%
4-Phenoxybenzenesulphonamide
Formel: C₁₂H₁₁NO₃S
Lagertemperatur: Raumtemperatur
MDL: MFCD00623227
CAS-Nummer: 123045-62-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR