1-Benzyl-3-pyrroline 96%

Lieferant: Apollo Scientific

OR480683-25G OR480683-5G
APOSOR480683-25GEA 0 EUR
APOSOR480683-25G APOSOR480683-5G
1-Benzyl-3-pyrroline 96%
1-Benzyl-3-pyrroline
Formel: C₁₁H₁₃N
Lagertemperatur: Raumtemperatur
MDL: MFCD00012324
CAS-Nummer: 6913-92-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR