N-(3-Aminophenyl)methanesulphonamide ≥95%

Lieferant: Apollo Scientific

Synonyme: 3-(Methylsulphonamido)aniline

OR480686-25G
APOSOR480686-25GEA 203 EUR
APOSOR480686-25G
N-(3-Aminophenyl)methanesulphonamide ≥95%
N-(3-Aminophenyl)methanesulphonamide
Formel: C₇H₁₀N₂O₂S
Lagertemperatur: Raumtemperatur
MDL: MFCD03094037
CAS-Nummer: 37045-73-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR