2-(4-Cyanophenoxy)-2-methylpropanoic acid 95%
Lieferant: Apollo Scientific
OR480687-1G
APOSOR480687-1GEA
105
EUR
APOSOR480687-1G
2-(4-Cyanophenoxy)-2-methylpropanoic acid 95%
2-(4-Cyanophenoxy)-2-methylpropanoic acid
Formel:
C₁₁H₁₁NO₃ Molecular Weight: 205,22 g/mol Lagertemperatur: Raumtemperatur |
MDL:
MFCD00084986 CAS-Nummer: 79925-16-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...