2,4-Dichlorobenzene-1,3-diol 95%

Lieferant: Apollo Scientific

OR480689-25G
APOSOR480689-25GEA 659 EUR
APOSOR480689-25G
2,4-Dichlorobenzene-1,3-diol 95%
2,4-Dichlorobenzene-1,3-diol
Formel: C₆H₄Cl₂O₂
Molecular Weight: 179,001 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD13152136
CAS-Nummer: 16606-61-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR