(4-Chloro-2,5-dimethyl-phenyl)-(4-methylsulphonylphenyl)methanone 95%

Lieferant: Apollo Scientific

OR480697-5G
APOSOR480697-5GEA 226 EUR
APOSOR480697-5G
(4-Chloro-2,5-dimethyl-phenyl)-(4-methylsulphonylphenyl)methanone 95%
(4-Chloro-2,5-dimethyl-phenyl)-(4-methylsulphonylphenyl)methanone
Formel: C₁₆H₁₅ClO₃S
Molecular Weight: 322,806 g/mol
Lagertemperatur: Raumtemperatur
MDL: MFCD21365058
CAS-Nummer: 1280729-47-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR