(R)-(+)-1,2-Epoxypropan 99%

Lieferant: Apollo Scientific

OR480701-25G
APOSOR480701-25GEA 236 EUR
APOSOR480701-25G
(R)-(+)-1,2-Epoxypropan 99%
(R)-(+)-1,2-Epoxypropan
Formel: C₃H₆O
Lagertemperatur: Raumtemperatur
MDL: MFCD00066211
CAS-Nummer: 15448-47-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR